COM448667798 Combi-Blocks, Inc.

2,5-Dihydrothiophene-3-carboxylic acid methyl ester

  • MDL number MFCD04038417
  • Cas Number : 67488-46-4
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6810

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6810
Availability details for
Quantity
  • Availability
  • Add to Cart