COM448667801 Combi-Blocks, Inc.

1-Bromo-2-methylhexadecane

  • MDL number MFCD00142588
  • Cas Number : 81367-59-1
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6853
Availability details for
Quantity
  • Availability
  • Add to Cart