COM448667803 Combi-Blocks, Inc.

Spirobicromane

  • MDL number MFCD00145320
  • Cas Number : 3127-14-8
  • Molecular Weight : 340.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6879
Availability details for
Quantity
  • Availability
  • Add to Cart