COM448667805 Combi-Blocks, Inc.

3-Methyltetrahydrofuran

  • MDL number MFCD00039681
  • Cas Number : 13423-15-9
  • Molecular Weight : 86.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6887
Availability details for
Quantity
  • Availability
  • Add to Cart