COM448667809 Combi-Blocks, Inc.

4,4'-Dibromo-4''-phenyltriphenylamine

  • MDL number MFCD08276360
  • Cas Number : 884530-69-2
  • Molecular Weight : 479.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6964
Availability details for
Quantity
  • Availability
  • Add to Cart