COM448667811 Combi-Blocks, Inc.

Trans-2,2-dimethyl-3-heptene

  • MDL number MFCD00039961
  • Cas Number : 19550-75-5
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7009
Availability details for
Quantity
  • Availability
  • Add to Cart