COM448667814 Combi-Blocks, Inc.

3-Hydroxyaspartic acid

  • MDL number MFCD00128126
  • Cas Number : 71653-06-0
  • Molecular Weight : 149.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7026
Availability details for
Quantity
  • Availability
  • Add to Cart