COM448667819 Combi-Blocks, Inc.

2,2,4-Trimethylcyclopentanone

  • MDL number MFCD00019312
  • Cas Number : 28056-54-4
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7120
Availability details for
Quantity
  • Availability
  • Add to Cart