COM448667828 Combi-Blocks, Inc.

4-Nitro-alpha,alpha,alpha',alpha'-tetrabromo-o-xylene

  • MDL number MFCD00143350
  • Cas Number : 13209-16-0
  • Molecular Weight : 466.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7229
Availability details for
Quantity
  • Availability
  • Add to Cart