COM448667830 Combi-Blocks, Inc.

N-Valeroyl bromide

  • MDL number MFCD00059467
  • Cas Number : 1889-26-5
  • Molecular Weight : 165.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7270
Availability details for
Quantity
  • Availability
  • Add to Cart