COM448667832 Combi-Blocks, Inc.

4,5-Dibromo-n-methoxy-n-methyl-2-furancarboxamide

  • MDL number MFCD08276337
  • Cas Number : 502639-02-3
  • Molecular Weight : 312.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7276
Availability details for
Quantity
  • Availability
  • Add to Cart