COM448667833 Combi-Blocks, Inc.

Bicyclo[3.2.1]-2-octene

  • MDL number MFCD00078236
  • Cas Number : 823-02-9
  • Molecular Weight : 108.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7286
Availability details for
Quantity
  • Availability
  • Add to Cart