COM448667843 Combi-Blocks, Inc.

2-(8-Chloro-1-naphthylthio)acetic acid

  • MDL number MFCD00043901
  • Cas Number : 129-94-2
  • Molecular Weight : 252.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7368
Availability details for
Quantity
  • Availability
  • Add to Cart