COM448667847 Combi-Blocks, Inc.

1-tert-Butyl-3-methylbenzene

  • MDL number MFCD00059210
  • Cas Number : 1075-38-3
  • Molecular Weight : 148.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7414
Availability details for
Quantity
  • Availability
  • Add to Cart