COM448667848 Combi-Blocks, Inc.

1,2-Nonanediol

  • MDL number MFCD01861291
  • Cas Number : 42789-13-9
  • Molecular Weight : 160.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7426
Availability details for
Quantity
  • Availability
  • Add to Cart