COM448667850 Combi-Blocks, Inc.

(R)-(+)-1-Ethyl-2-pyrrolidinecarboxylic acid ethyl ester

  • MDL number MFCD00671533
  • Cas Number : 381670-33-3
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7459
Availability details for
Quantity
  • Availability
  • Add to Cart