COM448667858 Combi-Blocks, Inc.

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate

  • MDL number MFCD00011943
  • Cas Number : 63212-53-3
  • Molecular Weight : 269.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7599
Availability details for
Quantity
  • Availability
  • Add to Cart