COM448667860 Combi-Blocks, Inc.

Alpha,alpha'-bis(4-methylbenzyloxy)-p-xylene

  • MDL number MFCD00143577
  • Cas Number : 136861-46-6
  • Molecular Weight : 346.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7626
Availability details for
Quantity
  • Availability
  • Add to Cart