COM448667870 Combi-Blocks, Inc.

1-Benzylpyrrole-3-carboxylic acid ethyl ester

  • MDL number MFCD06797056
  • Cas Number : 128259-47-2
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7718
Availability details for
Quantity
  • Availability
  • Add to Cart