COM448667874 Combi-Blocks, Inc.

1,2,9-Nonanetriol

  • MDL number MFCD01861292
  • Cas Number : 382631-42-7
  • Molecular Weight : 176.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7752
Availability details for
Quantity
  • Availability
  • Add to Cart