COM448667875 Combi-Blocks, Inc.

2-Naphthyl myristate

  • MDL number MFCD00037748
  • Cas Number : 7262-80-8
  • Molecular Weight : 354.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7755
Availability details for
Quantity
  • Availability
  • Add to Cart