COM448667876 Combi-Blocks, Inc.

2,4-Dimethylheptane

  • MDL number MFCD00048738
  • Cas Number : 2213-23-2
  • Molecular Weight : 128.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7757
Availability details for
Quantity
  • Availability
  • Add to Cart