COM448667877 Combi-Blocks, Inc.

1,5-Dimethyl-2-piperidone

  • MDL number MFCD03701098
  • Cas Number : 86917-58-0
  • Molecular Weight : 127.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7767
Availability details for
Quantity
  • Availability
  • Add to Cart