COM448667884 Combi-Blocks, Inc.

Tri-p-tolylbismuthine

  • MDL number MFCD00123272
  • Cas Number : 5142-75-6
  • Molecular Weight : 482.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7855
Availability details for
Quantity
  • Availability
  • Add to Cart