COM448667886 Combi-Blocks, Inc.

(Perfluoro-n-hexyl)phenyliodonium trifluoromethanesulfonate

  • MDL number MFCD00135119
  • Cas Number : 77758-84-0
  • Molecular Weight : 672.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7903
Availability details for
Quantity
  • Availability
  • Add to Cart