COM448667887 Combi-Blocks, Inc.

4-Methoxyphenyl 2,6-di-o-benzyl-beta-d-galactopyranoside

  • MDL number MFCD06797145
  • Cas Number : 159922-50-6
  • Molecular Weight : 466.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7907

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7907
Availability details for
Quantity
  • Availability
  • Add to Cart