COM448667888 Combi-Blocks, Inc.

N-Dodecylacrylamide

  • MDL number MFCD08276361
  • Cas Number : 1506-53-2
  • Molecular Weight : 239.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-7912
Availability details for
Quantity
  • Availability
  • Add to Cart