COM448667893 Combi-Blocks, Inc.

Cytidine sulfate

  • MDL number MFCD00151091
  • Cas Number : 32747-18-5
  • Molecular Weight : 341.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8034
Availability details for
Quantity
  • Availability
  • Add to Cart