COM448667904 Combi-Blocks, Inc.

2,2'-Diiodobiphenyl

  • MDL number MFCD00017622
  • Cas Number : 2236-52-4
  • Molecular Weight : 406.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8143
Availability details for
Quantity
  • Availability
  • Add to Cart