COM448667906 Combi-Blocks, Inc.

2-Methoxybutyl acetate

  • MDL number MFCD06797047
  • Cas Number : 1173168-18-7
  • Molecular Weight : 146.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8148
Availability details for
Quantity
  • Availability
  • Add to Cart