COM448667914 Combi-Blocks, Inc.

4-Ethoxyphenyl 4-butylbenzoate

  • MDL number MFCD00059426
  • Cas Number : 62716-65-8
  • Molecular Weight : 298.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8217
Availability details for
Quantity
  • Availability
  • Add to Cart