COM448667917 Combi-Blocks, Inc.

(1S,2R)-2-[N-Benzyl-n-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol

  • MDL number MFCD02093447
  • Cas Number : 187324-64-7
  • Molecular Weight : 423.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8227
Availability details for
Quantity
  • Availability
  • Add to Cart