COM448667920 Combi-Blocks, Inc.

(4As,8as)-2-benzoyl-1,3,4,4a,5,8a-hexahydro-6(2h)-isoquinolinone

  • MDL number MFCD06797067
  • Cas Number : 52346-14-2
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8237
Availability details for
Quantity
  • Availability
  • Add to Cart