COM448667926 Combi-Blocks, Inc.

(S,S)-Tadias-2nd

  • MDL number MFCD10566888
  • Molecular Weight : 972.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8312
Availability details for
Quantity
  • Availability
  • Add to Cart