COM448667935 Combi-Blocks, Inc.

N,2-Dimethyl-1,3-propanediamine

  • MDL number MFCD10566893
  • Cas Number : 1251384-75-4
  • Molecular Weight : 102.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8453
Availability details for
Quantity
  • Availability
  • Add to Cart