COM448667960 Combi-Blocks, Inc.

2,4,4-Trimethyl-1-hexene

  • MDL number MFCD00048626
  • Cas Number : 51174-12-0
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8778

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8778
Availability details for
Quantity
  • Availability
  • Add to Cart