COM448667965 Combi-Blocks, Inc.

(R)-(-)-2-Methylglutaric acid

  • MDL number MFCD00671562
  • Cas Number : 1115-81-7
  • Molecular Weight : 146.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8793

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8793
Availability details for
Quantity
  • Availability
  • Add to Cart