COM448667968 Combi-Blocks, Inc.

2,4-Dimethyl-1-heptene

  • MDL number MFCD00059250
  • Cas Number : 19549-87-2
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8807
Availability details for
Quantity
  • Availability
  • Add to Cart