COM448667970 Combi-Blocks, Inc.

2,5-Dibromofuran

  • MDL number MFCD00191342
  • Cas Number : 32460-00-7
  • Molecular Weight : 225.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8820

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-8820
Availability details for
Quantity
  • Availability
  • Add to Cart