COM448667983 Combi-Blocks, Inc.

3,6-Di(4-pyridyl)-1,2,4,5-tetrazine

  • MDL number MFCD00129054
  • Cas Number : 57654-36-1
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9067
Availability details for
Quantity
  • Availability
  • Add to Cart