COM448667986 Combi-Blocks, Inc.

5-Amino-6-chloro-2,3-dicyanopyrazine

  • MDL number MFCD00191417
  • Cas Number : 56413-96-8
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9129

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9129
Availability details for
Quantity
  • Availability
  • Add to Cart