COM448667995 Combi-Blocks, Inc.

Chloroacetyl-dl-isoleucine

  • MDL number MFCD00018911
  • Cas Number : 1115-24-8
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9251

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9251
Availability details for
Quantity
  • Availability
  • Add to Cart