COM448667999 Combi-Blocks, Inc.

1-Methylcyclopropane-1-carboxylic acid methyl ester

  • MDL number MFCD00191549
  • Cas Number : 6206-25-3
  • Molecular Weight : 114.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9292
Availability details for
Quantity
  • Availability
  • Add to Cart