COM448668003 Combi-Blocks, Inc.

Propionic acid (1r,2s)-2-[n-benzyl-n-(mesitylenesulfonyl)amino]-1-phenylpropyl ester

  • MDL number MFCD02093490
  • Cas Number : 187324-66-9
  • Molecular Weight : 479.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9359
Availability details for
Quantity
  • Availability
  • Add to Cart