COM448668011 Combi-Blocks, Inc.

2-Methyl-1,3-propanediamine

  • MDL number MFCD08276399
  • Cas Number : 2400-78-4
  • Molecular Weight : 88.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9484
Availability details for
Quantity
  • Availability
  • Add to Cart