COM448668016 Combi-Blocks, Inc.

4-Methoxyphenyl 3-o-allyl-4,6-o-benzylidene-2-deoxy-2-phthalimido-beta-d-glucopyranoside

  • MDL number MFCD08276388
  • Cas Number : 889453-84-3
  • Molecular Weight : 543.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9574
Availability details for
Quantity
  • Availability
  • Add to Cart