COM448668017 Combi-Blocks, Inc.

Ethyl undecafluoroamyl ketone

  • MDL number MFCD02093488
  • Cas Number : 383177-55-7
  • Molecular Weight : 326.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9593
Availability details for
Quantity
  • Availability
  • Add to Cart