COM448668024 Combi-Blocks, Inc.

4-Methoxyphenyl 3-o-benzyl-4,6-o-benzylidene-beta-d-glucopyranoside

  • MDL number MFCD08276394
  • Cas Number : 303127-81-3
  • Molecular Weight : 464.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9649
Availability details for
Quantity
  • Availability
  • Add to Cart