COM448668026 Combi-Blocks, Inc.

4-Methyl-2-hexene

  • MDL number MFCD00059242
  • Cas Number : 3404-55-5
  • Molecular Weight : 98.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9673

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9673
Availability details for
Quantity
  • Availability
  • Add to Cart