COM448668029 Combi-Blocks, Inc.

4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one

  • MDL number MFCD10566937
  • Cas Number : 632339-23-2
  • Molecular Weight : 154.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-9727
Availability details for
Quantity
  • Availability
  • Add to Cart